Learn More
ATHENA ES RPMI-1640 MEDIA WITHOUT ARG
Catalog No. NC0631685
Encompass_Preferred
SILAC media is prepared specifically for labeling experiments involving the use of stable amino acid isotopes. Athena provides several of the most commonly used media without L-arginine, L-lysine, L-leucine and L-methionine, the four mostly commonly used heavy isotope labeled amino acids.
Catalog No. NC0631685 Supplier ATHENA ES Supplier No. 0422
SILAC media is prepared specifically for labeling experiments involving the use of stable amino acid isotopes. Athena provides several of the most commonly used media without L-arginine, L-lysine, L-leucine and L-methionine, the four mostly commonly used heavy isotope labeled amino acids.