Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ RPMI-1640 Medium (1X) with 2.05 mM L-Glutamine

Catalog No. FB12999107 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FB12999107 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FB12999107

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ready-to-use basal medium that supports cell growth in research and biomanufacturing cell culture processes.

RPMI (Roswell Park Memorial Institute) 1640 Medium was originally developed to culture human leukemic cells. When supplemented with serum or a serum replacement, RPMI can be used with many cell types including human lymphocytes, Jurkat cells, HeLa cells, hybridomas and carcinomas.

This RPMI 1640 is modified with:

  • 2.05 mM L-Glutamine
TRUSTED_SUSTAINABILITY

Specifications

Cell Line Leukemic Cells
Cell Type HeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas
Concentration 1X
Form Liquid
Product Type RPMI 1640 Medium
Serum Level Standard Serum Supplementation
Sterility Sterile
Sterilization Method 0.1 μm Sterile-filtered
With Additives Glutamine
Without Additives None
Quantity 500 mL
Shipping Condition Ambient
Content And Storage 2°C to 8°C, away from light
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.