Learn More
TRIBIOSCIENCE INC RPMI-1640 Medium, 500 ML
SDP
Catalog No. NC3834042
Encompass
RPMI 1640 medium was originally formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of anchorage-dependent cells. It has also been shown to support several cell lines in the absence of serum.
Catalog No. NC3834042 Supplier TRIBIOSCIENCE INC Supplier No. TBS8063500ML
RPMI 1640 medium was originally formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of anchorage-dependent cells. It has also been shown to support several cell lines in the absence of serum.