Learn More
Sigma Aldrich Fine Chemicals Biosciences RPMI-1640 Medium HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture | MFCD00217820 | 500ML
Catalog No. NC2061693
Encompass
RPMI-1640 Medium HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture | MFCD00217820 | 500ML
Catalog No. NC2061693 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R5886500ML
RPMI-1640 Medium HEPES Modification, With 25 mM HEPES, without L-glutamine., liquid, sterile-filtered, suitable for cell culture | MFCD00217820 | 500ML