Learn More
Sigma Aldrich Fine Chemicals Biosciences RPMI-1640 Medium With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications | MFCD00217820 | 500ML
Catalog No. NC2340962
Encompass
RPMI-1640 Medium With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications | MFCD00217820 | 500ML
Catalog No. NC2340962 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R1780500ML
RPMI-1640 Medium With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications | MFCD00217820 | 500ML