Learn More
Sigma Aldrich Fine Chemicals Biosciences RPMI-1640 Medium With L-gl
Catalog No. 502900161
Encompass
RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoys 5A Medium it was formulated to support lymphoblastoid cells in suspension culture but it has since been shown to support a wide variety of cells that are anchorage dependent.
Catalog No. 50-290-0161 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R8758100ML
RPMI 1640 Medium was developed at Roswell Park Memorial Institute in 1966 by Moore and his co-workers. A modification of McCoys 5A Medium it was formulated to support lymphoblastoid cells in suspension culture but it has since been shown to support a wide variety of cells that are anchorage dependent.