Learn More
Sigma Aldrich Fine Chemicals Biosciences RPMI-1640 Medium With L-gl
Catalog No. 502901142
Encompass
Roswell Park Memorial Institute medium is commonly known as RPMI. It is a form of medium used in cell culture and tissue culture. This medium contains phosphate and is formulated for use in a 5% carbon dioxide atmosphere. It is used for the growth of human lymphoid cells.
Catalog No. 50-290-1142 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R650410L
Roswell Park Memorial Institute medium is commonly known as RPMI. It is a form of medium used in cell culture and tissue culture. This medium contains phosphate and is formulated for use in a 5% carbon dioxide atmosphere. It is used for the growth of human lymphoid cells.