Learn More
ILEX LIFE SCIENCES LLC RPMI 1640 P04-22100 500 ML
SDP
Catalog No. 502733804
Encompass_Preferred
PAN-Biotech RPMI 1640 Cell Culture Medium, w: L-Glutamine, w: 25 mM HEPES, w: 2.2 g/L NaHCO3 (6 x 500 ml). BICARBONATE: With 2.2 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: With L-Glutamine. PHENOL RED: With Phenol Red. HEPES: With 25 mM HEPES. SODIUM PYRUVATE: Without sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: RPMI 1640, also known as RPMI Medium, was originally developed at Roswell Park Memorial Institute (RPMI) in 1966 for the growth of human leukemia cells in monolayer or suspension cultures. RPMI media has since been found to be suitable for a wide variety of mammalian and hybridoma cells. We offer a variety of RPMI media to meet the needs of your specific applications.
Catalog No. 50-273-3804 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0422100
Add to Cart
Add to Cart
PAN-Biotech RPMI 1640 Cell Culture Medium, w: L-Glutamine, w: 25 mM HEPES, w: 2.2 g/L NaHCO3 (6 x 500 ml). BICARBONATE: With 2.2 g/L Sodium Bicarbonate. GLUCOSE: With glucose. GLUTAMINE: With L-Glutamine. PHENOL RED: With Phenol Red. HEPES: With 25 mM HEPES. SODIUM PYRUVATE: Without sodium pyruvate. STERILE: Yes. STORAGE TEMP: +2-8°C. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. DESCRIPTION: RPMI 1640, also known as RPMI Medium, was originally developed at Roswell Park Memorial Institute (RPMI) in 1966 for the growth of human leukemia cells in monolayer or suspension cultures. RPMI media has since been found to be suitable for a wide variety of mammalian and hybridoma cells. We offer a variety of RPMI media to meet the needs of your specific applications.