Learn More
Sigma Aldrich Fine Chemicals Biosciences RPMI 1640 Vitamins Solutio
Catalog No. 502901141
Encompass
Roswell Park Memorial Institute medium is commonly referred as RPMI. It is a form of medium used in cell culture and tissue culture. This medium contains phosphate and is formulated for use in a 5% carbon dioxide atmosphere. RPMI 1640 is used for the serum-free expansion of human lymphoid cells.
Catalog No. 50-290-1141 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R7256100ML
Roswell Park Memorial Institute medium is commonly referred as RPMI. It is a form of medium used in cell culture and tissue culture. This medium contains phosphate and is formulated for use in a 5% carbon dioxide atmosphere. RPMI 1640 is used for the serum-free expansion of human lymphoid cells.