Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc RPMI-1640 Medium with L-Glutamine, 500 mL
SDP

Catalog No. 50983285
Encompass_Preferred

Suitable for a wide range of anchorage independent cells. Commonly supplemented with 10% Fetal Bovine Serum (FBS). RPMI 1640 Medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. Use for fusion protocols and growth of hybrid cells, culture of normal human leukocytes in monolayer and suspension, and support of HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinoma growth (to name a few).

With 2.0 g/L Sodium Bicarbonate
Without HEPES buffer
With L-Glutamine
Storage Conditions: 2 °C to 8 °C
Shipping Condition: ambient temperature
Sterile Filtered pH: 7.0 ± 0.2
Osmolality: 300 ± 5% mOsM/Kg H2O
Max Shelf Life: 24 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-983-285 Supplier Quality Biological Inc Supplier No. 112025101
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Suitable for a wide range of anchorage independent cells. Commonly supplemented with 10% Fetal Bovine Serum (FBS). RPMI 1640 Medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. Use for fusion protocols and growth of hybrid cells, culture of normal human leukocytes in monolayer and suspension, and support of HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinoma growth (to name a few).

With 2.0 g/L Sodium Bicarbonate
Without HEPES buffer
With L-Glutamine
Storage Conditions: 2 °C to 8 °C
Shipping Condition: ambient temperature
Sterile Filtered pH: 7.0 ± 0.2
Osmolality: 300 ± 5% mOsM/Kg H2O
Max Shelf Life: 24 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.