Learn More
CYTION RPMI 8226 Cells, RPMI 8226 cells are a human myeloma cell line
Catalog No. NC3790633
Encompass
RPMI 8226 cells are a human myeloma cell line that was established in 1966 from the peripheral blood of a 61-year-old male patient with multiple myeloma. This cell line was named after the Roswell Park Memorial Institute (RPMI) where it was developed, and the number 8226 denotes its specific catalog number in the cell bank.The RPMI 8226 cell line is an important model system for studying multiple myeloma and related aspects of plasma cell biology, immunology research, and cancer therapy. RPMI 8226 cells are known to produce and secrete kappa light chains of immunoglobulins, a feature that is often exploited in research studies to investigate antibody production and secretion mechanisms.
Catalog No. NC3790633 Supplier CYTION Supplier No. 300431
RPMI 8226 cells are a human myeloma cell line that was established in 1966 from the peripheral blood of a 61-year-old male patient with multiple myeloma. This cell line was named after the Roswell Park Memorial Institute (RPMI) where it was developed, and the number 8226 denotes its specific catalog number in the cell bank.The RPMI 8226 cell line is an important model system for studying multiple myeloma and related aspects of plasma cell biology, immunology research, and cancer therapy. RPMI 8226 cells are known to produce and secrete kappa light chains of immunoglobulins, a feature that is often exploited in research studies to investigate antibody production and secretion mechanisms.