Learn More
ATCC RPMI 8226; Plasmacytoma; Human (Homo sapiens)
Supplier:  ATCC CCL155
Encompass
RPMI 8226 are B lymphocytes isolated in 1966 from the peripheral blood derived from a 61-year-old, male, plasmacytoma patient. RPMI 8226 can be used in immunology and immune system research.
Catalog No. 50-238-3647