Learn More
Sigma Aldrich Fine Chemicals Biosciences RPMI / F12 Complete Medium, with 2 mM L-Glut and 10% FBS |
Catalog No. NC1669043
Encompass
RPMI / F12 Complete Medium, with 2 mM L-Glut and 10% FBS |
Catalog No. NC1669043 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SLM242B
 Restricted Item
Add to Cart
Add to Cart
RPMI / F12 Complete Medium, with 2 mM L-Glut and 10% FBS |