Learn More
FUJIFILM Biosciences RPMI Medium 1640 1X HEPES
Catalog No. 5000450179
Encompass
RPMI Medium 1640 1X HEPES is designed for growth of humanLeukemia cells in either monolayers or suspension and is useful for a wide variety of suspension and monolayer cultures as well as for hybridomas
Catalog No. 50-004-50179 Supplier FUJIFILM Biosciences Supplier No. 9157500ML
RPMI Medium 1640 1X HEPES is designed for growth of humanLeukemia cells in either monolayers or suspension and is useful for a wide variety of suspension and monolayer cultures as well as for hybridomas