Learn More
FUJIFILM Biosciences RPMI Medium 1640 1X HEPES
Catalog No. 5000450180
Encompass
RPMI Medium 1640 1X HEPES w/oL-Glutamine is designed for growth of humanLeukemia cells in either monolayers or suspension and is useful for a wide variety of suspension and monolayer cultures as well as for hybridomas
Catalog No. 50-004-50180 Supplier FUJIFILM Biosciences Supplier No. 9159500ML
RPMI Medium 1640 1X HEPES w/oL-Glutamine is designed for growth of humanLeukemia cells in either monolayers or suspension and is useful for a wide variety of suspension and monolayer cultures as well as for hybridomas