Learn More
FUJIFILM BIOSCIENCES INC RPMI Medium 1640 1X HEPES
Supplier:  FUJIFILM BIOSCIENCES INC 9159100ML
Encompass
RPMI Medium 1640 1X HEPES w/oL-Glutamine is designed for growth of humanLeukemia cells in either monolayers or suspension and is useful for a wide variety of suspension and monolayer cultures as well as for hybridomas
Catalog No. 50-004-50249