Learn More
FUJIFILM Biosciences RPMI Medium 1640 w/oL-Glutamine - Liquid
Catalog No. NC9939886
Encompass
RPMI Medium 1640 w/oL-Glutamine is designed for growth of humanLeukemia cells in either monolayers or suspension and is useful for a wide variety of suspension and monolayer cultures, as well as for hybridomas.
Catalog No. NC9939886 Supplier FUJIFILM Biosciences Supplier No. 9160500ML
RPMI Medium 1640 w/oL-Glutamine is designed for growth of humanLeukemia cells in either monolayers or suspension and is useful for a wide variety of suspension and monolayer cultures, as well as for hybridomas.