Learn More
FUJIFILM Biosciences RPMI Medium 1640 - Liquid
Catalog No. 5000450255
Encompass
RPMI Medium 1640 is designed for growth of humanLeukemia cells in either monolayers or suspension and is useful for a wide variety of suspension and monolayer cultures as well as for hybridomas
Catalog No. 50-004-50255 Supplier FUJIFILM Biosciences Supplier No. 91611L
RPMI Medium 1640 is designed for growth of humanLeukemia cells in either monolayers or suspension and is useful for a wide variety of suspension and monolayer cultures as well as for hybridomas