Learn More
FUJIFILM BIOSCIENCES INC RPMI Medium 1640 - Liquid
Supplier:  FUJIFILM BIOSCIENCES INC 9161100ML
Encompass
RPMI Medium 1640 is designed for growth of humanLeukemia cells in either monolayers or suspension and is useful for a wide variety of suspension and monolayer cultures as well as for hybridomas
Catalog No. 50-004-50278