Learn More
FUJIFILM Biosciences RPMI Medium 1640 w/oL-Glutamin
Catalog No. 5000450183
Encompass
RPMI Medium 1640 w/oL-Glutamine is designed for growth of humanLeukemia cells in either monolayers or suspension and is useful for a wide variety of suspension and monolayer cultures as well as for hybridomas
Catalog No. 50-004-50183 Supplier FUJIFILM Biosciences Supplier No. 91601L
RPMI Medium 1640 w/oL-Glutamine is designed for growth of humanLeukemia cells in either monolayers or suspension and is useful for a wide variety of suspension and monolayer cultures as well as for hybridomas