Learn More
ILEX LIFE SCIENCES LLC PAN-Biotech RPMI 1640 Cell Culture Medium, w: L-Glutamine, w: 2.0 g/L NaHCO3 (500 ml)
SDP
Catalog No. 502388554
Encompass_Preferred
PAN-Biotech RPMI 1640 Cell Culture Medium, w: L-Glutamine, w: 2.0 g/L NaHCO3. QUANTITY: 500 ml. MANUFACTURED BY: Pan-Biotech (GERMANY). BICARBONATE: With 2.0 g/L Sodium Bicarbonate. GLUCOSE: with glucose. GLUTAMINE: with L-glutamine. HEPES: Without HEPES. LIQUID/POWDER: Liquid. PHENOL RED: With Phenol Red. SODIUM PYRUVATE: Without Sodium Pyruvate. STERILE: Yes. STORAGE TEMPERATURE: +2°C - +8°C. SUMMARY: RPMI 1640, also known as RPMI Medium, was originally developed at Roswell Park Memorial Institute (RPMI) in 1966 for the growth of human leukemia cells in monolayer or suspension cultures. RPMI media has since been found to be suitable for a wide variety of mammalian and hybridoma cells. We offer a variety of RPMI media to meet the needs of your specific applications.
Catalog No. 50-238-8554 Supplier ILEX LIFE SCIENCES LLC Supplier No. P0416500
PAN-Biotech RPMI 1640 Cell Culture Medium, w: L-Glutamine, w: 2.0 g/L NaHCO3. QUANTITY: 500 ml. MANUFACTURED BY: Pan-Biotech (GERMANY). BICARBONATE: With 2.0 g/L Sodium Bicarbonate. GLUCOSE: with glucose. GLUTAMINE: with L-glutamine. HEPES: Without HEPES. LIQUID/POWDER: Liquid. PHENOL RED: With Phenol Red. SODIUM PYRUVATE: Without Sodium Pyruvate. STERILE: Yes. STORAGE TEMPERATURE: +2°C - +8°C. SUMMARY: RPMI 1640, also known as RPMI Medium, was originally developed at Roswell Park Memorial Institute (RPMI) in 1966 for the growth of human leukemia cells in monolayer or suspension cultures. RPMI media has since been found to be suitable for a wide variety of mammalian and hybridoma cells. We offer a variety of RPMI media to meet the needs of your specific applications.