Learn More
Quality Biological Inc RPMI-1640 Medium with 25mM HEPES Buffer and L-Glutamine, 500 mL
SDP
Catalog No. 502209740
Encompass_Preferred
RPMI 1640 is suitable for a wide range of anchorage independent cells. RPMI 1640 Medium is commonly supplemented with 10% Fetal Bovine Serum (FBS). Although RPMI-1640 has been shown to support the serum-free growth of a number of cells.
With 2.0 g/L sodium bicarbonate
With HEPES buffer
Manufactured under an ISO 9001:2015 certified quality management system.
Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.
Catalog No. 50-220-9740 Supplier Quality Biological Inc Supplier No. 112143101
RPMI 1640 is suitable for a wide range of anchorage independent cells. RPMI 1640 Medium is commonly supplemented with 10% Fetal Bovine Serum (FBS). Although RPMI-1640 has been shown to support the serum-free growth of a number of cells.
With 2.0 g/L sodium bicarbonate
With HEPES buffer
Manufactured under an ISO 9001:2015 certified quality management system.
Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.