Learn More
Quality Biological Inc RPMI-1640 Medium with Glutamine XL™, Case 10 X 500 mL
SDP
Catalog No. 502209745
Encompass_Preferred
RRPMI 1640 is suitable for a wide range of anchorage independent cells. RPMI 1640 Medium is commonly supplemented with 10% Fetal Bovine Serum (FBS). Although RPMI-1640 has been shown to support the serum-free growth of a number of cells.
With 2.0 g/L sodium bicarbonate
Without HEPES buffer
With stable L-analyl-glutamine (Glutamine XL)
Manufactured under an ISO 9001:2015 certified quality management system.
Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.
Catalog No. 50-220-9745 Supplier Quality Biological Inc Supplier No. 112301101CS
RRPMI 1640 is suitable for a wide range of anchorage independent cells. RPMI 1640 Medium is commonly supplemented with 10% Fetal Bovine Serum (FBS). Although RPMI-1640 has been shown to support the serum-free growth of a number of cells.
With 2.0 g/L sodium bicarbonate
Without HEPES buffer
With stable L-analyl-glutamine (Glutamine XL)
Manufactured under an ISO 9001:2015 certified quality management system.
Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.