Learn More
ATCC RPMI-1640 Medium
Catalog No. 502383277
Encompass
RPMI-1640 medium modified to contain 2 mM L-glutamine, 10 mM HEPES, 1 mM sodium pyruvate, 4500 mg/L glucose, and 1500 mg/L sodium bicarbonate, for use in incubators using 5% CO2 in air. Additional sodium bicarbonate may be required for use in incubators containing higher percentages of CO2. It has applications for cell culture, cell growth, and viability.
Catalog No. 50-238-3277 Supplier ATCC Supplier No. 302001
RPMI-1640 medium modified to contain 2 mM L-glutamine, 10 mM HEPES, 1 mM sodium pyruvate, 4500 mg/L glucose, and 1500 mg/L sodium bicarbonate, for use in incubators using 5% CO2 in air. Additional sodium bicarbonate may be required for use in incubators containing higher percentages of CO2. It has applications for cell culture, cell growth, and viability.