Learn More
Aviva Systems Biology Corp RPP30 Antibody - C-terminal region (ARP64629_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64629_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPP30. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human RPP30. Alternate names: FLJ38491, TSG15ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Rabbit
Catalog No. 50-288-319