Learn More
Aviva Systems Biology Corp RPP40 Antibody - N-terminal region (ARP62358_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62358_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPP40. It was validated on Western Blot. . Alternate names: RNASEP1, bA428J1.3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-287-659