Learn More
Medchemexpress LLC RPR132595A hydrochloride | 185304-42-1 | >98.0% | 518.6 g/mol | C28H30N4O6 | 5 MG
SDP
Catalog No. 5000214252
Encompass_Preferred
RPR132595A hydrochloride is the hydrochloride salt of RPR132595A (NPC), an active metabolite of the anticancer prodrug irinotecan (CPT-11). It is formed by cytochrome P450 3A4 (CYP3A4) metabolism and is supplied as a high-purity research reagent for analytical and biochemical studies.
  • High purity (>98%) suitable for analytical and biochemical assays.
  • Provided in small, accurately quantified quantities for reference studies.
  • Represents the primary active metabolite of irinotecan formed by CYP3A4.
  • Soluble in DMSO and common organic solvents for sample preparation.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-14252 Supplier Medchemexpress LLC Supplier No. HY100638A5MG
RPR132595A hydrochloride is the hydrochloride salt of RPR132595A (NPC), an active metabolite of the anticancer prodrug irinotecan (CPT-11). It is formed by cytochrome P450 3A4 (CYP3A4) metabolism and is supplied as a high-purity research reagent for analytical and biochemical studies.
  • High purity (>98%) suitable for analytical and biochemical assays.
  • Provided in small, accurately quantified quantities for reference studies.
  • Represents the primary active metabolite of irinotecan formed by CYP3A4.
  • Soluble in DMSO and common organic solvents for sample preparation.
  • Intended for research use only; not for human or veterinary use.