Learn More
Cayman Chemical RPramIpexolehydrochLRId 10mg
SDP
Catalog No. 502966464
Encompass_Preferred
An enantiomer of pramipexole that is ~100-fold less active than the (S) form as a dopamine receptor agonist; an antioxidant that targets mitochondria to prevent apoptosis
Catalog No. 50-296-6464 Supplier Cayman Chemical Supplier No. 1823410 MG
An enantiomer of pramipexole that is ~100-fold less active than the (S) form as a dopamine receptor agonist; an antioxidant that targets mitochondria to prevent apoptosis