Learn More
Cayman Chemical (R)-Pramipexole (hydrochloride), Purity: ≥95%, CAS: 104632-27-1, 50 mg
SDP
Catalog No. 502966460
Encompass_Preferred
An enantiomer of pramipexole that is ~100-fold less active than the (S) form as a dopamine receptor agonist; an antioxidant that targets mitochondria to prevent apoptosis
Catalog No. 50-296-6460 Supplier Cayman Chemical Supplier No. 1823450 MG
An enantiomer of pramipexole that is ~100-fold less active than the (S) form as a dopamine receptor agonist; an antioxidant that targets mitochondria to prevent apoptosis