Learn More
Aviva Systems Biology Corp RPRD1A Antibody - N-terminal region (ARP62702_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62702_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPRD1A. It was validated on Western Blot. . Alternate names: FLJ10656, HsT3101, MGC19513, P15RSApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-747