Learn More
Aviva Systems Biology Corp RPRD1B Antibody - middle region (ARP57534_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57534_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against C20orf77. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RPRD1B. Alternate names: CREPT, DKFZp434P0735, FLJ44520, dJ1057B20.2, NET60, C20orf77ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53876