Learn More
Sigma Aldrich Fine Chemicals Biosciences rProtein A GraviTrapTM Cyt
Catalog No. 502908003
Encompass
rProtein A GraviTrap(TM) includes ten ready-to-use columns prepacked with 1 mL of rProtein A Sepharose(R) Fast Flow.
Catalog No. 50-290-8003 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE28985254
rProtein A GraviTrap(TM) includes ten ready-to-use columns prepacked with 1 mL of rProtein A Sepharose(R) Fast Flow.