Learn More
Aviva Systems Biology Corp RPS11 Antibody - N-terminal region (ARP65144_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65144_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPS11. It was validated on Western Blot. . Alternate names: S11ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-288-441