Learn More
Aviva Systems Biology Corp RPS13 Antibody - middle region (ARP56180_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56180_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPS13. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RPS13. Alternate names: S13ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-286-123