Learn More
Aviva Systems Biology Corp RPS14 Antibody - N-terminal region (ARP40321_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40321_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RPS14. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RPS14. Alternate names: S14, EMTBApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57391