Learn More
Aviva Systems Biology Corp RPS16 Antibody - middle region (ARP33536_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33536_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RPS16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RPS16. Alternate names: S16ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55058