Learn More
Aviva Systems Biology Corp RPS17 Antibody - middle region (ARP64463_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64463_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPS17. It was validated on Western Blot. . Alternate names: DBA4, MGC72007, RPS17L1, RPS17L2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-288-268