Learn More
Aviva Systems Biology Corp RPS2 Antibody - middle region (ARP63572_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63572_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPS2. It was validated on Western Blot. . Alternate names: LLREP3, MGC102851, MGC117344, MGC117345, S2ApplicationWB. Species reactivity: Dog, Pig, Rat, Yeast, Zebrafish
Catalog No. 50-288-016