Learn More
Aviva Systems Biology Corp RPS21 Antibody - N-terminal region (ARP56208_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56208_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPS21. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RPS21. Alternate names: S21ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-61758