Learn More
Aviva Systems Biology Corp RPS24 Antibody - middle region (ARP40304_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40304_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPS24. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RPS24. Alternate names: DBA3, DKFZp686N1586, S24ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57386