Learn More
Aviva Systems Biology Corp RPS27A Antibody - middle region (ARP59416_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59416_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPS27A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RPS27A. Alternate names: CEP80, UBA80, UBCEP1, UBCEP80, UBC, S27AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast, Zebrafish
Catalog No. 50-286-690