Learn More
Aviva Systems Biology Corp RPS27L Antibody - N-terminal region (ARP56775_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56775_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPS27L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RPS27L. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-61985