Learn More
Aviva Systems Biology Corp RPS29 Antibody - N-terminal region (ARP40329_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40329_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against RPS29. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RPS29. Alternate names: S29ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-53190