Learn More
Aviva Systems Biology Corp RPS3 Antibody - C-terminal region (ARP63708_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63708_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPS3. It was validated on Western Blot. . Alternate names: FLJ26283, FLJ27450, MGC87870, S3ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-288-058