Learn More
Aviva Systems Biology Corp RPS3A Antibody - N-terminal region (ARP56138_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56138_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPS3A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RPS3A. Alternate names: FTE1, MFTL, MGC23240, S3AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61735