Learn More
Aviva Systems Biology Corp RPS4Y1 Antibody - C-terminal region (ARP62231_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62231_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPS4Y1. It was validated on Western Blot. . Alternate names: MGC119100, MGC5070, RPS4Y, S4ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-622