Learn More
Enzo Life Sciences Ribosomal protein S6 (rat) (229-239) (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP1440001
Encompass_Preferred
This sequence, AKRRRLSSLRA, derived from rat hepatic 40S ribosomal protein S6 (229-239). Substrate for Rho-kinase II (ROCK II), Km=0.9 µM and for p90 rsk (S6K) in cell extracts (Km=0.18 mM) using phosphocellulose assay. Peptide phosphorylation by PKS and PKC can be eliminated by the inclusion of PKI and EGTA, respectively, in the assay. Long Term Storage: -20°C.
Catalog No. 50-201-1254