Learn More
Aviva Systems Biology Corp RPS6KA2 Antibody - middle region (ARP56170_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56170_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RPS6KA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RPS6KA2. Alternate names: HU-2, MAPKAPK1C, RSK, RSK3, S6K-alpha, S6K-alpha2, p90-RSK3, pp90RSK3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61745