Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human RPS6KA2 (RSK3), GST Tag Recombinant Protein

Catalog No. A31518
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A31518 100 μg
A31522 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A31518 Supplier Invitrogen™ Supplier No. A31518
Add to Cart
Add to Cart

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. The purified recombinant full-length protein can be used in a number of different applications, but it has been specifically validated for use with both the LanthaScreen™ & Z'-LYTE™ Assay technologies to identify novel kinase inhibitors. RPS6KA2 is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the LanthaScreen & Z'-LYTE assay technology Available in multiple pack sizes for convenient use Also available in our SelectSceen Kinase Profiling Service Protein Form: Full Length, Recombinant, Active The gene accession number for RPS6KA2 (RSK3) is NP_066958.2.

RPS6KA2 (RSK3) serine/threonine kinase is a member of the ribosomal S6 kinase (RSK) family. It is believed that RPS6KA2 is a signal-transducing intermediate in the cellular response to several growth factors. It is found in many tissues, but in higher levels in lung and skeletal muscle. The p90 ribosomal S6 kinases (RSK)1-4 are downstream members of the extracellular signal-regulated kinase (ERK)/MAPK cascade. The loss of RSK2 activity in humans leads to Coffin-Lowry syndrome, which is characterized by mental retardation and growth deficits and may have a tonic regulation on G-protein coupled signaling.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15349
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol, 10mM glutathione, 150mM NaCl and no preservative; pH 7.5
Gene ID (Entrez) 6196
Molecular Weight (g/mol) 109.9 kDa
Name Human RPS6KA2 (RSK3), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 100 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 90 kDa ribosomal protein S6 kinase 2; 90kDa; D17Wsu134e; HU-2; MAP kinase-activated protein kinase 1c; MAPK-activated protein kinase 1c; MAPKAP kinase 1C; MAPKAPK1C; MAPKAPK-1c; mitogen-activated protein kinase-activated protein kinase 1C; p90rsk; p90-RSK 2; p90RSK2; p90-RSK3; pp90rsk; pp90RSK3; Protein-tyrosine kinase Mpk-9; ribosomal protein S6 kinase A2; ribosomal protein S6 kinase alpha 2; ribosomal protein S6 kinase alpha-2; ribosomal protein S6 kinase polypeptide 2; ribosomal protein S6 kinase, 90kD, polypeptide 2; ribosomal protein S6 kinase, 90kDa, polypeptide 2; ribosomal protein S6 kinase, polypeptide 2; ribosomal S6 kinase 3; Rps6ka2; Rps6ka-rs1; RSK; Rsk3; RSK-3; S6K-alpha; S6K-alpha 2; S6K-alpha2; S6K-alpha-2
Common Name RSK3
Gene Symbol RPS6KA2
Kinase Family RSK (RPS6K) Kinases
Protein Length Full Length
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol.

Store at -80°C. Upon first use, aliquot and store at -80°C to avoid multiple freeze/thaws.
Shipping Condition Dry Ice
Expression System Insect cells
Protein Form Full Length, Recombinant, Active
Form Liquid
Research Category Drug Development
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.