Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human RPS6KA4 (MSK2), GST Tag Recombinant Protein

Catalog No. PV3782
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV3782 10 μg
PR7155A 100 μg
PV4052 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PV3782 Supplier Invitrogen™ Supplier No. PV3782
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Full Length.

RPS6KA4 (MSK2) is a serine/threonine protein kinase that is essential for the regulation of various cellular processes. This kinase contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including CREB1 and c-fos. RPS6KA4 contains 2 non-identical kinase catalytic domains and phosphorylates various substrates, including CREB1 and c-fos. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75676
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 0.02% Triton X-100, 50% glycerol, 2mM DTT, 0.5mM EDTA, 150mM NaCl and no preservative; pH 7.5
Gene ID (Entrez) 8986
Molecular Weight (g/mol) 113.1 kDa
Name Human RPS6KA4 (MSK2), GST Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 1110069D02Rik; 90 kDa ribosomal protein S6 kinase 4; 90kDa; AI848992; Ccdc88b; coiled-coil domain containing 88B; EC 2.7.11.1; kinase MSK2; KS6A4; mitogen- and stress-activated protein kinase 2; mitogen- and stress-activated protein kinase-2; mMSK2; MSK2; Nuclear mitogen- and stress-activated protein kinase 2; ribosomal protein kinase B; ribosomal protein S6 kinase A4; ribosomal protein S6 kinase alpha 4; ribosomal protein S6 kinase alpha-4; ribosomal protein S6 kinase, 90kD, polypeptide 4; ribosomal protein S6 kinase, 90kDa, polypeptide 4; ribosomal protein S6 kinase, polypeptide 4; RLSK; RPS6KA4; RSKB; RSK-B; RSK-like protein kinase; S6K-alpha-4
Common Name MSK2
Gene Symbol Rps6ka4
Kinase Family RSK (RPS6K) Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Kinase Group Ser⁄Thr Kinases: AGC Group
Shipping Condition Dry Ice
System Compatibility LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™
Target Entry RPS6KA4
Target Gene ribosomal protein S6 kinase, 90 kDa, polypeptide 4
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.